Assay Details
Target Gene Details
Entrez Gene ID: | 25861 |
Gene Name: | whirlin |
Gene Aliases: |
CIP98, DFNB31, PDZD7B, USH2D, WI |
Location: |
Chr.9:114402078-114505509 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WHRN | NM_001083885.2 | 9 | 1348 | NP_001077354.2 |
| NM_001173425.1 | 9 | 2512 | NP_001166896.1 | |
| NM_015404.3 | 9 | 2512 | NP_056219.3 | |
| XM_011518484.1 | 10 | 2598 | XP_011516786.1 | |
| XM_011518485.1 | 11 | 2446 | XP_011516787.1 | |
| XM_011518486.1 | 10 | 2598 | XP_011516788.1 | |
| XM_011518487.1 | 9 | 2472 | XP_011516789.1 | |
| XM_011518488.1 | 8 | 2355 | XP_011516790.1 | |
| XM_011518495.1 | 7 | 850 | XP_011516797.1 | |
| AB040959.1 | 9 | 2026 | ||
| AK022854.1 | 5 | 1226 | BAB14275.1 | |
| AL110228.1 | 9 | 1306 | CAB53685.2 | |
| BC136416.1 | 9 | 2261 | ||
| BC142614.1 | 9 | 2512 | ||
| BC142684.1 | 9 | 2487 | ||
| BX329269.1 | 4 | 722 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv948n27 | Chr.9:114193988 - 114693895 on Build GRCh38 | Gain |
|
| dgv7706n100 | Chr.9:114182745 - 114697870 on Build GRCh38 | Gain |
|
| nsv831693 | Chr.9:114354170 - 114519615 on Build GRCh38 | Gain |
|
| dgv33e196 | Chr.9:114030664 - 114604363 on Build GRCh38 | Duplication |
|
| nsv615233 | Chr.9:114167820 - 114486199 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map