Assay Details
Target Gene Details
Entrez Gene ID: | 7297 |
Gene Name: | tyrosine kinase 2 |
Gene Aliases: |
IMD35, JTK1 |
Location: |
Chr.19:10350528-10380572 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TYK2 | NM_003331.4 | 13 | 2287 | NP_003322.3 |
| XM_011528245.1 | 13 | 2197 | XP_011526547.1 | |
| XM_011528246.2 | 12 | 2022 | XP_011526548.1 | |
| XM_011528247.1 | 11 | 1799 | XP_011526549.1 | |
| XM_011528249.2 | 5 | 745 | XP_011526551.1 | |
| XM_011528251.1 | XP_011526553.1 | |||
| XM_011528252.1 | XP_011526554.1 | |||
| AK222672.1 | 13 | 2240 | BAD96392.1 | |
| AK303627.1 | 8 | 1357 | ||
| AK308360.1 | 9 | 1502 | ||
| AK308379.1 | 4 | 319 | ||
| BC014243.2 | 13 | 2250 | AAH14243.1 | |
| X54637.1 | 13 | 2215 | CAA38449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518008 | Chr.19:10338682 - 10426592 on Build GRCh38 | Gain |
|
| nsv2415 | Chr.19:10340631 - 10373197 on Build GRCh38 | Insertion |
|
| nsv470124 | Chr.19:10289228 - 10362257 on Build GRCh38 | Gain |
|
| nsv470125 | Chr.19:10334150 - 10380799 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map