Assay Details
Target Gene Details
Entrez Gene ID: | 8886 |
Gene Name: | DEAD-box helicase 18 |
Gene Aliases: |
MrDb |
Location: |
Chr.2:117814679-117832377 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX18 | NM_006773.3 | NP_006764.3 | ||
| AK001467.1 | BAA91709.1 | |||
| BC003360.1 | AAH03360.1 | |||
| BC024739.1 | AAH24739.1 | |||
| BU599536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999434 | Chr.2:117074067 - 117975257 on Build GRCh38 | Gain |
|
| nsv2889 | Chr.2:117811935 - 117840276 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map