Assay Details
Target Gene Details
Entrez Gene ID: | 3588 |
Gene Name: | interleukin 10 receptor subunit beta |
Gene Aliases: |
CDW210B, CRF2-4, CRFB4, D21S58, D21S66, IL-10R2 |
Location: |
Chr.21:33266360-33297234 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL10RB | NM_000628.4 | 3 | 371 | NP_000619.3 |
| AK124057.1 | 2 | 682 | BAC85761.1 | |
| AK299961.1 | 3 | 272 | ||
| AK313348.1 | 3 | 364 | ||
| AU133530.2 | 3 | 371 | ||
| BC001903.1 | 3 | 350 | AAH01903.1 | |
| BT009777.1 | 3 | 265 | AAP88779.1 | |
| JN591378.1 | 3 | 265 | ||
| Z17227.1 | 3 | 308 | CAA78933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587376 | Chr.21:33271777 - 33307426 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map