Assay Details
Target Gene Details
Entrez Gene ID: | 6259 |
Gene Name: | receptor-like tyrosine kinase |
Gene Aliases: |
D3S3195, JTK5, JTK5A, RYK1 |
Location: |
Chr.3:134157133-134250834 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RYK | NM_001005861.2 | 15 | 1957 | NP_001005861.1 |
| NM_002958.3 | 15 | 1948 | NP_002949.2 | |
| XM_017007016.1 | 10 | 1454 | XP_016862505.1 | |
| AB209405.1 | 5 | 4284 | BAD92642.1 | |
| BC021700.2 | 10 | 1223 | AAH21700.2 | |
| S59184.1 | 15 | 1987 | AAB26341.1 | |
| X69970.1 | 15 | 1948 | CAA49591.1 | |
| X96588.1 | 16 | 1853 | CAA65406.1 |
More Information
Set Membership: |
|

Genomic Map