Assay Details
Target Gene Details
Entrez Gene ID: | 220972 |
Gene Name: | membrane associated ring-CH-type finger 8 |
Gene Aliases: |
CMIR, MARCH-VIII, MIR, RNF178, c-MIR |
Location: |
Chr.10:45454585-45594907 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARCH8 | NM_001002266.2 | 7 | 1443 | NP_001002266.1 |
| NM_001282866.1 | 8 | 2289 | NP_001269795.1 | |
| NM_145021.5 | 7 | 1931 | NP_659458.2 | |
| XM_005271804.2 | 8 | 2778 | XP_005271861.1 | |
| XM_006717704.2 | 8 | 2193 | XP_006717767.1 | |
| XM_011539492.2 | 9 | 2829 | XP_011537794.1 | |
| XM_011539493.2 | 10 | 3070 | XP_011537795.1 | |
| XM_011539494.2 | 9 | 3019 | XP_011537796.1 | |
| XM_011539495.1 | 7 | 1327 | XP_011537797.1 | |
| XM_017015894.1 | 8 | 1983 | XP_016871383.1 | |
| AK023636.1 | 3 | 1732 | ||
| AK057289.1 | 7 | 1637 | ||
| AL833316.1 | 4 | 1674 | ||
| BC025394.2 | 7 | 1420 | AAH25394.1 | |
| BC066988.1 | 7 | 1932 | AAH66988.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043536 | Chr.10:45420493 - 45580787 on Build GRCh38 | Gain |
|
| esv3891797 | Chr.10:45008026 - 45674466 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map