Assay Details
Target Gene Details
Entrez Gene ID: | 22890 |
Gene Name: | zinc finger and BTB domain containing 1 |
Gene Aliases: |
ZNF909 |
Location: |
Chr.14:64504574-64533690 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB1 | NM_001123329.1 | NP_001116801.1 | ||
| NM_014950.2 | NP_055765.2 | |||
| XM_005267410.1 | XP_005267467.1 | |||
| XM_011536566.2 | XP_011534868.1 | |||
| XM_011536568.2 | XP_011534870.1 | |||
| XM_017021095.1 | XP_016876584.1 | |||
| AB023214.1 | ||||
| AK291743.1 | ||||
| BC050719.1 | AAH50719.1 | |||
| BX248777.1 | CAD66584.1 |
Target Gene Details
Entrez Gene ID: | 7597 |
Gene Name: | zinc finger and BTB domain containing 25 |
Gene Aliases: |
C14orf51, KUP, ZNF46 |
Location: |
Chr.14:64447172-64505213 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB25 | NM_001304507.1 | 1 | 282 | NP_001291436.1 |
| NM_001304508.1 | 1 | 282 | NP_001291437.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1126366 | Chr.14:64503682 - 64505482 on Build GRCh38 | Deletion |
|
| nsv521823 | Chr.14:64502624 - 64528976 on Build GRCh38 | Loss |
|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map