Assay Details
Target Gene Details
Entrez Gene ID: | 338323 |
Gene Name: | NLR family pyrin domain containing 14 |
Gene Aliases: |
CLR11.2, GC-LRR, NALP14, NOD5, PAN8 |
Location: |
Chr.11:7020446-7089025 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP14 | NM_176822.3 | NP_789792.1 | ||
| XM_011520044.1 | XP_011518346.1 | |||
| AK310900.1 | ||||
| AY154469.1 | AAO18165.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832061 | Chr.11:6847937 - 7049841 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map