Assay Details
Target Gene Details
Entrez Gene ID: | 1237 |
Gene Name: | C-C motif chemokine receptor 8 |
Gene Aliases: |
CC-CKR-8, CCR-8, CDw198, CKRL1, CMKBR8, CMKBRL2, CY6, GPRCY6, TER1 |
Location: |
Chr.3:39329706-39333680 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCR8 | NM_005201.3 | NP_005192.1 | ||
| AF005210.1 | AAB62547.1 | |||
| AK314957.1 | ||||
| BC107152.2 | AAI07153.1 | |||
| BC107159.1 | AAI07160.1 | |||
| D49919.1 | BAA23387.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3595925 | Chr.3:39314483 - 39353966 on Build GRCh38 | Loss |
|
| esv29441 | Chr.3:39309994 - 39372588 on Build GRCh38 | Loss |
|
| nsv460509 | Chr.3:39315814 - 39356217 on Build GRCh38 | Loss |
|
| nsv518801 | Chr.3:39296450 - 39356217 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map