Assay Details
Target Gene Details
Entrez Gene ID: | 7318 |
Gene Name: | ubiquitin like modifier activating enzyme 7 |
Gene Aliases: |
D8, UBA1B, UBE1L, UBE2 |
Location: |
Chr.3:49805205-49813958 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA7 | NM_003335.2 | 13 | 1733 | NP_003326.2 |
| XM_005265430.2 | 13 | 1717 | XP_005265487.1 | |
| XM_006713321.2 | XP_006713384.1 | |||
| XM_011534070.1 | 12 | 1561 | XP_011532372.1 | |
| XM_011534071.1 | XP_011532373.1 | |||
| XM_011534072.1 | 12 | 1626 | XP_011532374.1 | |
| BC006378.2 | 13 | 1658 | AAH06378.1 | |
| BT007026.1 | 13 | 1562 | AAP35672.1 | |
| L13852.1 | 13 | 1723 | AAA75388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834688 | Chr.3:49773682 - 49981866 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map