Assay Details
Target Gene Details
Entrez Gene ID: | 286223 |
Gene Name: | chromosome 9 open reading frame 47 |
Gene Aliases: |
C9orf108, bA791O21.3 |
Location: |
Chr.9:88990863-88996142 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C9orf47 | NM_001001938.3 | 3 | 2549 | NP_001001938.1 |
| NM_001142413.1 | 3 | 2492 | NP_001135885.1 | |
| AK127854.1 | 3 | 2485 | BAC87162.1 | |
| AK130650.1 | ||||
| BC136895.1 | 3 | 2382 | ||
| BC144432.1 | 3 | 2383 |
Target Gene Details
Entrez Gene ID: | 1903 |
Gene Name: | sphingosine-1-phosphate receptor 3 |
Gene Aliases: |
EDG-3, EDG3, LPB3, S1P3 |
Location: |
Chr.9:88991409-89005155 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S1PR3 | NM_005226.3 | NP_005217.2 | ||
| AK312798.1 | ||||
| BC060827.1 | AAH60827.1 | |||
| DA722994.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621014 | Chr.9:88877891 - 89641453 on Build GRCh38 | Gain |
|
| esv29340 | Chr.9:88907171 - 89010602 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map