Assay Details
Target Gene Details
Entrez Gene ID: | 2232 |
Gene Name: | ferredoxin reductase |
Gene Aliases: |
ADXR |
Location: |
Chr.17:74862497-74873031 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss |
|
| nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss |
|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map