Assay Details
Target Gene Details
Entrez Gene ID: | 102465513 |
Gene Name: | microRNA 6852 |
Gene Aliases: |
hsa-mir-6852 |
Location: |
Chr.9:35710676-35710741 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6852 | NR_106911.1 |
Target Gene Details
Entrez Gene ID: | 7094 |
Gene Name: | talin 1 |
Gene Aliases: |
ILWEQ, TLN |
Location: |
Chr.9:35697337-35732395 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 32 - Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLN1 | NM_006289.3 | NP_006280.3 | ||
| AB028950.2 | ||||
| AF078828.1 | AAD13152.1 | |||
| AF177198.1 | AAF23322.1 | |||
| BC042923.1 | AAH42923.1 |
More Information
Set Membership: |
|

Genomic Map