Assay Details
Target Gene Details
Entrez Gene ID: | 285464 |
Gene Name: | cysteine rich PAK1 inhibitor |
Gene Aliases: |
- |
Location: |
Chr.4:1391552-1395994 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRIPAK | NM_175918.3 | 1 | 4219 | NP_787114.2 |
| AA160784.1 | 1 | 249 | ||
| AK091762.1 | 1 | 1726 | BAC03741.1 | |
| AK096209.1 | 1 | 1407 | BAC04725.1 | |
| AK126919.1 | 1 | 4219 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss |
|
| esv21856 | Chr.4:1394190 - 1398850 on Build GRCh38 | Gain+Loss |
|
| nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss |
|
| nsv4197 | Chr.4:1387665 - 1421471 on Build GRCh38 | Insertion |
|
| nsv593302 | Chr.4:1344830 - 1420896 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map