Assay Details
Target Gene Details
Entrez Gene ID: | 84527 |
Gene Name: | zinc finger protein 559 |
Gene Aliases: |
NBLA00121 |
Location: |
Chr.19:9323772-9343845 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF559 | NM_001202406.1 | NP_001189335.1 | ||
| NM_001202407.1 | NP_001189336.1 | |||
| NM_001202408.1 | NP_001189337.1 | |||
| NM_001202409.1 | NP_001189338.1 | |||
| NM_001202410.1 | NP_001189339.1 | |||
| NM_001202411.1 | NP_001189340.1 | |||
| NM_001202412.1 | NP_001189341.1 | |||
| NM_032497.2 | NP_115886.1 | |||
| AK027700.1 | ||||
| AK056489.1 | ||||
| AK092348.1 | ||||
| AK298169.1 | ||||
| AK308517.1 | ||||
| AK309952.1 | ||||
| AK310020.1 | ||||
| AK310117.1 | ||||
| AK310781.1 | ||||
| AL389938.1 | ||||
| BC006436.1 | AAH06436.1 | |||
| BP337457.1 | ||||
| BP417021.1 | ||||
| CF597362.1 | ||||
| DA509657.1 | ||||
| DA517887.1 | ||||
| DA731101.1 | ||||
| DA743493.1 | ||||
| DA774704.1 | ||||
| DB158737.1 | ||||
| DB288007.1 |
Target Gene Details
Entrez Gene ID: | 100529215 |
Gene Name: | ZNF559-ZNF177 readthrough |
Gene Aliases: |
- |
Location: |
Chr.19:9324226-9382617 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF559-ZNF177 | NM_001172650.2 | NP_001166121.1 | ||
| NM_001202425.1 | NP_001189354.1 | |||
| U37251.1 | AAB09748.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819521 | Chr.19:9338335 - 9338764 on Build GRCh38 | Gain |
|
| esv2674742 | Chr.19:9331339 - 9340550 on Build GRCh38 | Deletion |
|
| nsv523472 | Chr.19:9180514 - 9340901 on Build GRCh38 | Loss |
|
| nsv521091 | Chr.19:9290483 - 9339353 on Build GRCh38 | Loss |
|
| esv3643633 | Chr.19:9331314 - 9340641 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map