Assay Details
Target Gene Details
Entrez Gene ID: | 8467 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5 |
Gene Aliases: |
ISWI, SNF2H, WCRF135, hISWI, hSNF2H |
Location: |
Chr.4:143513463-143557489 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMARCA5 | NM_003601.3 | NP_003592.3 | ||
| AB010882.1 | BAA25173.1 | |||
| AK302841.1 | ||||
| AK308424.1 | ||||
| AK316444.1 | ||||
| BC023144.1 | AAH23144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759288 | Chr.4:143427140 - 144210439 on Build GRCh38 | Gain+Loss |
|
| nsv830103 | Chr.4:143472245 - 143631892 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map