Assay Details
Target Gene Details
Entrez Gene ID: | 51691 |
Gene Name: | LSM8 homolog, U6 small nuclear RNA associated |
Gene Aliases: |
NAA38 |
Location: |
Chr.7:118184032-118204039 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LSM8 | NM_016200.4 | 4 | 434 | NP_057284.1 |
| AF182294.1 | 4 | 324 | AAD56232.1 | |
| AK311936.1 | ||||
| BC002742.2 | 4 | 280 | AAH02742.1 | |
| BC022440.1 | 4 | 304 | AAH22440.1 | |
| DA112078.1 | 4 | 303 | ||
| DA318705.1 | 4 | 401 | ||
| DA478457.1 | 4 | 302 | ||
| DB275674.1 | 4 | 435 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608259 | Chr.7:117837974 - 119521689 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map