Assay Details
Target Gene Details
Entrez Gene ID: | 79772 |
Gene Name: | multiple C2 and transmembrane domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.5:94703690-95284575 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 28 - Exon 28 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv598953 | Chr.5:94621232 - 94721120 on Build GRCh38 | Loss |
|
| esv3570255 | Chr.5:94703088 - 94719414 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map