Assay Details
Target Gene Details
Entrez Gene ID: | 101927027 |
Gene Name: | uncharacterized LOC101927027 |
Gene Aliases: |
- |
Location: |
Chr.2:178413659-178440243 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927027 | NR_110204.1 | |||
| AL520497.3 | ||||
| BX371397.2 | ||||
| BX538254.1 | 5 | 2599 |
Target Gene Details
Entrez Gene ID: | 8575 |
Gene Name: | protein activator of interferon induced protein kinase EIF2AK2 |
Gene Aliases: |
DYT16, HSD14, PACT, RAX |
Location: |
Chr.2:178431414-178451240 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKRA | NM_001139517.1 | 6 | 946 | NP_001132989.1 |
| NM_001139518.1 | 7 | 667 | NP_001132990.1 | |
| NM_001316362.1 | 8 | 985 | NP_001303291.1 | |
| NM_003690.4 | 7 | 866 | NP_003681.1 | |
| XM_011512063.1 | 7 | 797 | XP_011510365.1 | |
| XM_011512066.1 | 8 | 1022 | XP_011510368.1 | |
| XM_017005159.1 | 8 | 907 | XP_016860648.1 | |
| AF072860.1 | 7 | 761 | AAC25672.1 | |
| AF083033.1 | 7 | 757 | AAD33099.1 | |
| AK093360.1 | 3 | 892 | ||
| AK223107.1 | 7 | 765 | BAD96827.1 | |
| AK290601.1 | 7 | 792 | ||
| AK296019.1 | 8 | 896 | ||
| AL136615.1 | 7 | 790 | CAB66550.1 | |
| AL833867.1 | 6 | 946 | CAD38725.1 | |
| AY251164.1 | 7 | 684 | AAP20061.1 | |
| BC009470.1 | 7 | 772 | AAH09470.1 | |
| BT007243.1 | 7 | 665 | AAP35907.1 | |
| CR533525.1 | 7 | 665 | CAG38556.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv625e201 | Chr.2:178432240 - 178441593 on Build GRCh38 | Deletion |
|
| esv8989 | Chr.2:178432002 - 178436300 on Build GRCh38 | Loss |
|
| nsv834467 | Chr.2:178393413 - 178566238 on Build GRCh38 | Loss |
|
| esv7371 | Chr.2:178436147 - 178443360 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map