Assay Details
Target Gene Details
Entrez Gene ID: | 23542 |
Gene Name: | mitogen-activated protein kinase 8 interacting protein 2 |
Gene Aliases: |
IB-2, IB2, JIP2, PRKM8IPL |
Location: |
Chr.22:50600685-50613978 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK8IP2 | NM_012324.4 | 5 | 1204 | NP_036456.1 |
| XM_011530679.2 | 5 | 1096 | XP_011528981.1 | |
| XM_011530680.2 | 5 | 982 | XP_011528982.1 | |
| XM_011530681.2 | 5 | 961 | XP_011528983.1 | |
| AF136382.1 | 5 | 1187 | AAF00980.1 | |
| AF218778.1 | 3 | 989 | AAF32323.1 | |
| AK293580.1 | ||||
| AK294265.1 | ||||
| AL021708.1 | 1 | 207 | CAA16714.1 | |
| BC009940.2 | AAH09940.2 | |||
| BC047527.1 | 5 | 1117 | AAH47527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589224 | Chr.22:50589773 - 50639823 on Build GRCh38 | Loss |
|
| nsv1059234 | Chr.22:50560544 - 50617472 on Build GRCh38 | Gain |
|
| nsv1067172 | Chr.22:50586195 - 50677098 on Build GRCh38 | Gain |
|
| nsv829343 | Chr.22:50599349 - 50606643 on Build GRCh38 | Gain |
|
| nsv834239 | Chr.22:50485317 - 50630215 on Build GRCh38 | Loss |
|
| nsv955203 | Chr.22:50599072 - 50631372 on Build GRCh38 | Deletion |
|
| nsv589223 | Chr.22:50539595 - 50619495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map