Assay Details
Target Gene Details
Entrez Gene ID: | 1381 |
Gene Name: | cellular retinoic acid binding protein 1 |
Gene Aliases: |
CRABP, CRABP-I, CRABPI, RBP5 |
Location: |
Chr.15:78340324-78348230 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRABP1 | NM_004378.2 | NP_004369.1 | ||
| AB593091.1 | ||||
| AI863257.1 | ||||
| AL110113.1 | ||||
| AU132726.1 | ||||
| BC022069.1 | AAH22069.1 | |||
| CR457017.1 | CAG33298.1 | |||
| S74445.1 | AAB20773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052910 | Chr.15:78316295 - 78414707 on Build GRCh38 | Gain |
|
| nsv833063 | Chr.15:78312873 - 78462603 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs151297065] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map