Assay Details
Target Gene Details
Entrez Gene ID: | 109 |
Gene Name: | adenylate cyclase 3 |
Gene Aliases: |
AC-III, AC3 |
Location: |
Chr.2:24819169-24920017 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY3 | NM_001320613.1 | 22 | 4515 | NP_001307542.1 |
| NM_004036.4 | 22 | 4512 | NP_004027.2 | |
| XM_006711925.1 | 23 | 4297 | XP_006711988.1 | |
| XM_011532489.1 | 23 | 4354 | XP_011530791.1 | |
| XM_011532490.1 | 23 | 4351 | XP_011530792.1 | |
| XM_011532491.1 | 22 | 4288 | XP_011530793.1 | |
| XM_011532492.2 | 24 | 4422 | XP_011530794.1 | |
| XM_011532493.1 | 23 | 4216 | XP_011530795.1 | |
| XM_011532494.1 | 23 | 4156 | XP_011530796.1 | |
| XM_011532496.1 | 22 | 3521 | XP_011530798.1 | |
| XM_017003186.1 | 23 | 4294 | XP_016858675.1 | |
| XM_017003187.1 | 22 | 4285 | XP_016858676.1 | |
| XM_017003188.1 | 24 | 4419 | XP_016858677.1 | |
| XM_017003189.1 | 23 | 4213 | XP_016858678.1 | |
| XM_017003190.1 | 22 | 4090 | XP_016858679.1 | |
| XM_017003191.1 | 22 | 3562 | XP_016858680.1 | |
| XM_017003192.1 | 21 | 3398 | XP_016858681.1 | |
| XM_017003193.1 | 21 | 3395 | XP_016858682.1 | |
| AB011083.1 | 21 | 3348 | BAA25437.1 | |
| AF033861.1 | AAD13403.1 | |||
| AK027857.1 | 21 | 3861 | ||
| AK075424.1 | 9 | 1723 | BAC11613.1 | |
| AK095173.1 | 19 | 2921 | ||
| AK122926.1 | 22 | 4110 |
Target Gene Details
Entrez Gene ID: | 79172 |
Gene Name: | centromere protein O |
Gene Aliases: |
CENP-O, ICEN-36, MCM21R |
Location: |
Chr.2:24793115-24822376 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPO | NM_001199803.2 | NP_001186732.1 | ||
| NM_001322101.1 | NP_001309030.1 | |||
| NM_024322.3 | NP_077298.1 | |||
| NR_136182.1 | ||||
| NR_136183.1 | ||||
| NR_136184.1 | ||||
| AK027859.1 | BAB55414.1 | |||
| BC002870.1 | AAH02870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004529 | Chr.2:24363242 - 25005468 on Build GRCh38 | Loss |
|
| nsv508831 | Chr.2:24787088 - 24857561 on Build GRCh38 | Insertion |
|
| nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain |
|
| nsv1000020 | Chr.2:24709475 - 24909903 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map