Assay Details
Target Gene Details
Entrez Gene ID: | 114609 |
Gene Name: | TIR domain containing adaptor protein |
Gene Aliases: |
BACTS1, Mal, MyD88-2, wyatt |
Location: |
Chr.11:126281268-126294933 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIRAP | NM_001039661.1 | NP_001034750.1 | ||
| NM_001318776.1 | 4 | 922 | NP_001305705.1 | |
| NM_001318777.1 | NP_001305706.1 | |||
| NM_148910.2 | 5 | 1091 | NP_683708.1 | |
| XM_005271399.3 | 3 | 820 | XP_005271456.2 | |
| XM_011542576.2 | XP_011540878.1 | |||
| XM_017017161.1 | XP_016872650.1 | |||
| XM_017017162.1 | XP_016872651.1 | |||
| AB446477.1 | ||||
| AF378129.1 | 5 | 1072 | AAL05627.1 | |
| AF406652.1 | 2 | 639 | AAL01160.1 | |
| AF410783.1 | AAL05036.1 | |||
| AK124298.1 | ||||
| AK313147.1 | 4 | 915 | ||
| AY576785.1 | AAT90417.1 | |||
| AY576786.1 | AAT90418.1 | |||
| AY576787.1 | 2 | 678 | AAT90419.1 | |
| BC032474.1 | AAH32474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556530 | Chr.11:126264724 - 126393990 on Build GRCh38 | Loss |
|
| esv2762950 | Chr.11:126290159 - 126304652 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map