Assay Details
Target Gene Details
Entrez Gene ID: | 146862 |
Gene Name: | unc-45 myosin chaperone B |
Gene Aliases: |
CMYA4, CTRCT43, SMUNC45, UNC-45B, UNC45 |
Location: |
Chr.17:35147793-35189345 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC45B | NM_001033576.1 | NP_001028748.1 | ||
| NM_001267052.1 | NP_001253981.1 | |||
| XM_017024234.1 | XP_016879723.1 | |||
| AK095929.1 | ||||
| AL832355.1 | ||||
| AL833281.1 | ||||
| BX647642.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892998 | Chr.17:35106468 - 35295755 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map