Assay Details
Target Gene Details
Entrez Gene ID: | 8552 |
Gene Name: | inactivation escape 1 (non-protein coding) |
Gene Aliases: |
NCRNA00010 |
Location: |
Chr.X:47204848-47205861 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INE1 | NR_024616.1 | 1 | 605 | |
| Y10696.1 | 1 | 532 | CAA71702.2 |
Target Gene Details
Entrez Gene ID: | 7317 |
Gene Name: | ubiquitin like modifier activating enzyme 1 |
Gene Aliases: |
A1S9, A1S9T, A1ST, AMCX1, CFAP124, GXP1, POC20, SMAX2, UBA1A, UBE1, UBE1X |
Location: |
Chr.X:47190800-47215128 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA1 | NM_003334.3 | NP_003325.2 | ||
| NM_153280.2 | NP_695012.1 | |||
| XM_005272649.1 | XP_005272706.1 | |||
| XM_011543954.2 | XP_011542256.1 | |||
| XM_017029777.1 | XP_016885266.1 | |||
| XM_017029778.1 | XP_016885267.1 | |||
| XM_017029779.1 | XP_016885268.1 | |||
| XM_017029780.1 | XP_016885269.1 | |||
| XM_017029781.1 | XP_016885270.1 | |||
| AK293161.1 | ||||
| AK296866.1 | ||||
| BC013041.2 | AAH13041.1 | |||
| M58028.1 | AAA61246.1 | |||
| X52897.1 | CAA37078.1 | |||
| X56976.1 | CAA40296.1 |
More Information
Set Membership: |
|

Genomic Map