Assay Details
Target Gene Details
Entrez Gene ID: | 200014 |
Gene Name: | coiled-coil and C2 domain containing 1B |
Gene Aliases: |
- |
Location: |
Chr.1:52348437-52366291 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CC2D1B | NM_032449.2 | NP_115825.1 | ||
| XM_005270590.2 | XP_005270647.1 | |||
| XM_005270591.2 | XP_005270648.1 | |||
| XM_005270593.2 | XP_005270650.1 | |||
| XM_006710423.2 | XP_006710486.1 | |||
| XM_017000559.1 | XP_016856048.1 | |||
| AB058739.1 | BAB47465.1 | |||
| AK130874.1 | BAC85450.1 | |||
| BC007912.2 | AAH07912.2 | |||
| BC016880.1 | AAH16880.1 | |||
| BC039308.1 | AAH39308.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950728 | Chr.1:52351829 - 52363128 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map