Assay Details
Target Gene Details
Entrez Gene ID: | 116986 |
Gene Name: | ArfGAP with GTPase domain, ankyrin repeat and PH domain 2 |
Gene Aliases: |
CENTG1, GGAP2, PIKE |
Location: |
Chr.12:57723765-57742201 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGAP2 | NM_001122772.2 | 19 | 3961 | NP_001116244.1 |
| NM_014770.3 | 18 | 2983 | NP_055585.1 | |
| XM_005268625.3 | 18 | 4027 | XP_005268682.1 | |
| XM_005268626.2 | 19 | 3083 | XP_005268683.1 | |
| XM_017018772.1 | 19 | 4088 | XP_016874261.1 | |
| AF384128.1 | 18 | 2979 | AAO39848.1 | |
| D79989.1 | 18 | 2981 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470296 | Chr.12:57718406 - 57783509 on Build GRCh38 | Loss |
|
| nsv826383 | Chr.12:57724297 - 57729864 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map