Assay Details
Target Gene Details
Entrez Gene ID: | 53833 |
Gene Name: | interleukin 20 receptor subunit beta |
Gene Aliases: |
DIRS1, FNDC6, IL-20R2 |
Location: |
Chr.3:136957865-137011085 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL20RB | NM_144717.3 | 6 | 981 | NP_653318.2 |
| XM_006713665.3 | 6 | 1710 | XP_006713728.1 | |
| XM_011512910.2 | 7 | 885 | XP_011511212.1 | |
| XM_011512911.2 | 6 | 838 | XP_011511213.1 | |
| XM_017006632.1 | 5 | 794 | XP_016862121.1 | |
| AK296832.1 | 6 | 853 | ||
| AK316084.1 | 6 | 861 | ||
| AY358305.1 | 6 | 981 | AAQ88672.1 | |
| BC033292.1 | 5 | 821 | AAH33292.1 | |
| BC063696.1 | 6 | 845 | AAH63696.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597895 | Chr.3:136981487 - 137109810 on Build GRCh38 | Gain |
|
| esv3597894 | Chr.3:136980610 - 137047680 on Build GRCh38 | Gain |
|
| nsv524856 | Chr.3:136974617 - 137471923 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map