Assay Details
Target Gene Details
Entrez Gene ID: | 23283 |
Gene Name: | cleavage stimulation factor subunit 2, tau variant |
Gene Aliases: |
CstF-64T |
Location: |
Chr.10:51695486-51699595 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSTF2T | NM_015235.2 | 1 | 2949 | NP_056050.1 |
| AB014589.1 | 1 | 2697 | BAA31664.1 |
Target Gene Details
Entrez Gene ID: | 5592 |
Gene Name: | protein kinase, cGMP-dependent, type I |
Gene Aliases: |
AAT8, PKG, PRKG1B, PRKGR1B, cGK, cGK 1, cGK1, cGKI, cGKI-BETA, cGKI-alpha |
Location: |
Chr.10:50991151-52298350 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKG1 | NM_001098512.2 | NP_001091982.1 | ||
| NM_006258.3 | NP_006249.1 | |||
| XM_011539952.2 | XP_011538254.1 | |||
| XM_017016412.1 | XP_016871901.1 | |||
| XM_017016413.1 | XP_016871902.1 | |||
| AK313416.1 | ||||
| BC062688.1 | AAH62688.1 | |||
| BC127090.1 | ||||
| D45864.1 | BAA08297.1 | |||
| EF560730.1 | ||||
| Y07512.1 | CAA68810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045692 | Chr.10:51541690 - 52327063 on Build GRCh38 | Gain |
|
| nsv831875 | Chr.10:51595665 - 51789367 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map