Assay Details
Target Gene Details
Entrez Gene ID: | 55783 |
Gene Name: | cap methyltransferase 2 |
Gene Aliases: |
AFT, FTSJD1, HMTr2, MTr2 |
Location: |
Chr.16:71282300-71290178 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTR2 | NM_001099642.1 | 3 | 1840 | NP_001093112.1 |
| NM_001324374.1 | 3 | 1771 | NP_001311303.1 | |
| NM_001324377.1 | 3 | 1542 | NP_001311306.1 | |
| NM_001324378.1 | 2 | 1795 | NP_001311307.1 | |
| NM_001324379.1 | 2 | 1857 | NP_001311308.1 | |
| NM_018348.5 | 3 | 1984 | NP_060818.4 | |
| XM_011523228.1 | 3 | 2129 | XP_011521530.1 | |
| XM_011523229.1 | 3 | 2124 | XP_011521531.1 | |
| XM_017023444.1 | 3 | 2327 | XP_016878933.1 | |
| AK000289.1 | 1 | 373 | BAA91058.1 | |
| AK002033.1 | 1 | 1568 | BAA92047.1 | |
| AK023183.1 | 1 | 1273 | BAB14452.1 | |
| AK226169.1 | 3 | 1735 | ||
| AK315056.1 | 3 | 1782 | ||
| BC034468.1 | 3 | 1566 | AAH34468.1 | |
| BC035005.2 | 3 | 1771 | AAH35005.1 | |
| BX648660.1 | 3 | 1747 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509631 | Chr.16:71279342 - 71336612 on Build GRCh38 | Insertion |
|
| nsv1861 | Chr.16:71279772 - 71298716 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map