Assay Details
Target Gene Details
Entrez Gene ID: | 9138 |
Gene Name: | Rho guanine nucleotide exchange factor 1 |
Gene Aliases: |
GEF1, LBCL2, LSC, P115-RHOGEF, SUB1.5 |
Location: |
Chr.19:41883200-41907454 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 17 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF1 | NM_004706.3 | NP_004697.2 | ||
| NM_198977.1 | NP_945328.1 | |||
| NM_199002.1 | NP_945353.1 | |||
| XM_005259386.4 | XP_005259443.1 | |||
| XM_005259390.4 | XP_005259447.1 | |||
| XM_011527468.2 | XP_011525770.1 | |||
| AL832294.1 | ||||
| BC005155.2 | AAH05155.2 | |||
| BC011726.2 | AAH11726.1 | |||
| BC034013.1 | AAH34013.2 | |||
| BC035127.1 | AAH35127.1 | |||
| BC067262.1 | AAH67262.1 | |||
| BT007421.1 | AAP36089.1 | |||
| U64105.1 | AAB17896.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv953580 | Chr.19:41888230 - 41997948 on Build GRCh38 | Deletion |
|
| nsv579625 | Chr.19:41896804 - 41902357 on Build GRCh38 | Loss |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map