Assay Details
Target Gene Details
Entrez Gene ID: | 79048 |
Gene Name: | SECIS binding protein 2 |
Gene Aliases: |
SBP2 |
Location: |
Chr.9:89318473-89367117 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621014 | Chr.9:88877891 - 89641453 on Build GRCh38 | Gain |
|
| nsv1041327 | Chr.9:89298100 - 89438209 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map