Assay Details
Target Gene Details
Entrez Gene ID: | 2232 |
Gene Name: | ferredoxin reductase |
Gene Aliases: |
ADXR |
Location: |
Chr.17:74862497-74873031 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FDXR | NM_001258012.3 | 12 | 1883 | NP_001244941.2 |
| NM_001258013.3 | 13 | 1847 | NP_001244942.2 | |
| NM_001258014.3 | 12 | 1730 | NP_001244943.2 | |
| NM_001258015.2 | 11 | 1634 | NP_001244944.1 | |
| NM_001258016.3 | 11 | 1699 | NP_001244945.2 | |
| NM_004110.5 | 12 | 1772 | NP_004101.3 | |
| NM_024417.4 | 12 | 1754 | NP_077728.3 | |
| NR_047576.2 | 12 | 1854 | ||
| XM_006721772.1 | 12 | 1702 | XP_006721835.1 | |
| XM_011524527.1 | 12 | 1720 | XP_011522829.1 | |
| XM_011524528.1 | 12 | 1809 | XP_011522830.1 | |
| XM_011524529.1 | 12 | 1812 | XP_011522831.1 | |
| XM_011524531.2 | 11 | 1705 | XP_011522833.1 | |
| XM_011524532.2 | 11 | 1712 | XP_011522834.1 | |
| XM_011524533.2 | 11 | 1680 | XP_011522835.1 | |
| AK094120.1 | 12 | 2032 | ||
| AK098648.1 | 12 | 1691 | ||
| AK223530.1 | BAD97250.1 | |||
| AK293208.1 | ||||
| AK293210.1 | 12 | 1791 | ||
| AK295307.1 | 12 | 1883 | ||
| AK298908.1 | 11 | 1577 | ||
| AK301779.1 | 12 | 1667 | ||
| AK301977.1 | 11 | 1699 | ||
| AW590330.1 | 1 | 141 | ||
| BC002960.1 | 12 | 1693 | AAH02960.1 | |
| BC063493.1 | 12 | 1692 | AAH63493.1 | |
| J03826.1 | 12 | 1705 | AAB59497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss |
|
| nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss |
|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map