Assay Details
Target Gene Details
Entrez Gene ID: | 22807 |
Gene Name: | IKAROS family zinc finger 2 |
Gene Aliases: |
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2 |
Location: |
Chr.2:212999684-213152753 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834528 | Chr.2:212799464 - 213014337 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv834529 | Chr.2:212886244 - 213093670 on Build GRCh38 | Gain |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
