Assay Details
Target Gene Details
Entrez Gene ID: | 23019 |
Gene Name: | CCR4-NOT transcription complex subunit 1 |
Gene Aliases: |
AD-005, CDC39, NOT1, NOT1H |
Location: |
Chr.16:58519946-58629886 on Build GRCh38 |
Assay Gene Location: | Within Exon 44 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT1 | NM_001265612.1 | 44 | 6642 | NP_001252541.1 |
| NM_016284.4 | 44 | 6657 | NP_057368.3 | |
| NR_049763.1 | 44 | 6642 | ||
| AB023224.2 | 31 | 4702 | BAA76851.2 | |
| AF110778.1 | 4 | 434 | AAF14861.1 | |
| AK056927.1 | 21 | 2987 | ||
| AK075125.1 | 14 | 1956 | ||
| AK092885.1 | 4 | 1201 | ||
| AK301092.1 | 4 | 421 | ||
| AK302495.1 | 21 | 3027 | ||
| AL117492.1 | 16 | 2496 | CAB55960.1 | |
| AL832769.1 | 44 | 6606 | ||
| AL833549.1 | 44 | 6616 | ||
| BC000779.2 | 6 | 815 | AAH00779.2 | |
| BC024317.1 | 27 | 4056 | AAH24317.1 | |
| BX537840.1 | 44 | 6588 | CAD97851.1 | |
| CR749237.1 | 37 | 5597 | CAH18093.1 | |
| EF553522.1 | 44 | 6591 |
More Information
Set Membership: |
|

Genomic Map