Assay Details
Target Gene Details
Entrez Gene ID: | 51564 |
Gene Name: | histone deacetylase 7 |
Gene Aliases: |
HD7, HD7A, HDAC7A |
Location: |
Chr.12:47782710-47820612 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC7 | NM_001098416.3 | 16 | 2127 | NP_001091886.1 |
| NM_001308090.1 | 16 | 2187 | NP_001295019.1 | |
| NM_015401.4 | 17 | 2238 | NP_056216.2 | |
| XM_011538478.1 | 17 | 2190 | XP_011536780.1 | |
| XM_011538479.1 | 18 | 2632 | XP_011536781.1 | |
| XM_011538480.1 | 16 | 2079 | XP_011536782.1 | |
| XM_011538481.1 | 17 | 2083 | XP_011536783.1 | |
| XM_011538482.1 | 17 | 2167 | XP_011536784.1 | |
| XM_017019455.1 | 16 | 2139 | XP_016874944.1 | |
| XM_017019456.1 | XP_016874945.1 | |||
| AF239243.1 | 14 | 2021 | AAF63491.1 | |
| AK001032.1 | BAA91474.1 | |||
| AK001190.1 | 9 | 1160 | BAA91545.1 | |
| AK024469.1 | 12 | 2218 | BAB15759.1 | |
| AK122588.1 | 3 | 3196 | BAC56929.1 | |
| AK128383.1 | 9 | 1081 | ||
| AK225910.1 | ||||
| AK299292.1 | 17 | 2153 | ||
| AK301545.1 | 17 | 2101 | ||
| AK303481.1 | 16 | 2093 | ||
| AL117455.1 | 13 | 1705 | CAB55935.1 | |
| AY302468.1 | 17 | 2043 | AAQ18232.1 | |
| AY321367.1 | 16 | 2012 | AAP84704.1 | |
| BC006453.1 | 6 | 662 | ||
| BC020505.2 | 9 | 1205 | AAH20505.2 | |
| BC064840.1 | 16 | 2021 | AAH64840.1 | |
| BT009771.1 | 7 | 912 | AAP88773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv94n21 | Chr.12:47725779 - 47821397 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map