Assay Details
Target Gene Details
Entrez Gene ID: | 121457 |
Gene Name: | IKBKB interacting protein |
Gene Aliases: |
IKIP |
Location: |
Chr.12:98613404-98645051 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKBIP | NM_153687.3 | NP_710154.1 | ||
| NM_201612.2 | NP_963906.1 | |||
| NM_201613.2 | NP_963907.1 | |||
| AJ539425.1 | CAD62380.1 | |||
| AJ539426.1 | CAD62381.1 | |||
| AJ539427.1 | CAD62382.1 | |||
| AK055613.1 | BAB70970.1 | |||
| AK123069.1 | BAC85528.1 | |||
| BC029415.1 | ||||
| BC051372.1 | ||||
| BC058933.1 | AAH58933.1 | |||
| BC107572.1 | AAI07573.1 | |||
| BM468651.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819940 | Chr.12:98626516 - 98634052 on Build GRCh38 | Loss |
|
| nsv559918 | Chr.12:98570959 - 98666572 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map