Assay Details
Target Gene Details
Entrez Gene ID: | 7328 |
Gene Name: | ubiquitin conjugating enzyme E2 H |
Gene Aliases: |
E2-20K, GID3, UBC8, UBCH, UBCH2 |
Location: |
Chr.7:129830732-129952960 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2H | NM_001202498.1 | NP_001189427.1 | ||
| NM_003344.3 | NP_003335.1 | |||
| NM_182697.2 | NP_874356.1 | |||
| AB451490.1 | ||||
| AK091088.1 | 1 | 818 | ||
| AK316590.1 | ||||
| AL832212.1 | 1 | 890 | ||
| AY302138.1 | AAP57630.1 | |||
| BC006277.2 | AAH06277.1 | |||
| BI088146.1 | ||||
| BT006756.1 | AAP35402.1 | |||
| KJ534959.1 | ||||
| Z29328.1 | CAA82525.1 | |||
| Z29330.1 | CAA82527.1 | |||
| Z29331.1 | CAA82528.1 |
More Information
Set Membership: |
|

Genomic Map