Assay Details
Target Gene Details
Entrez Gene ID: | 4661 |
Gene Name: | myelin transcription factor 1 |
Gene Aliases: |
C20orf36, MTF1, MYTI, NZF2, PLPB1, ZC2H2C1, ZC2HC4A |
Location: |
Chr.20:64164474-64242253 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYT1 | NM_004535.2 | NP_004526.1 | ||
| AB020642.1 | ||||
| BC062313.1 | AAH62313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470564 | Chr.20:64027492 - 64169828 on Build GRCh38 | Gain |
|
| nsv428381 | Chr.20:63583375 - 64334167 on Build GRCh38 | Gain+Loss |
|
| nsv524962 | Chr.20:64141284 - 64247978 on Build GRCh38 | Loss |
|
| dgv4347n100 | Chr.20:64131041 - 64324788 on Build GRCh38 | Gain |
|
| esv2758807 | Chr.20:64006869 - 64334167 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map