Assay Details
Target Gene Details
Entrez Gene ID: | 134637 |
Gene Name: | adenosine deaminase, tRNA specific 2 |
Gene Aliases: |
DEADC1, TAD2, dJ20N2, dJ20N2.1 |
Location: |
Chr.6:143422832-143450750 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAT2 | NM_001286259.1 | NP_001273188.1 | ||
| NM_182503.2 | NP_872309.2 | |||
| XM_011535439.2 | XP_011533741.1 | |||
| XM_011535440.1 | XP_011533742.1 | |||
| XM_011535444.2 | XP_011533746.1 | |||
| XM_017010260.1 | XP_016865749.1 | |||
| XM_017010261.1 | XP_016865750.1 | |||
| XM_017010262.1 | XP_016865751.1 | |||
| XM_017010263.1 | XP_016865752.1 | |||
| AK126201.1 | ||||
| BC037955.1 | ||||
| BX538182.1 | CAD98054.1 |
More Information
Set Membership: |
|

Genomic Map