Assay Details
Target Gene Details
Entrez Gene ID: | 64651 |
Gene Name: | cysteine and serine rich nuclear protein 1 |
Gene Aliases: |
AXUD1, CSRNP-1, FAM130B, TAIP-3, URAX1 |
Location: |
Chr.3:39141851-39154602 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSRNP1 | NM_001320559.1 | 2 | 404 | NP_001307488.1 |
| NM_001320560.1 | 2 | 205 | NP_001307489.1 | |
| NM_033027.3 | 2 | 273 | NP_149016.2 | |
| XM_017007048.1 | 2 | 3369 | XP_016862537.1 | |
| AB053121.1 | 2 | 250 | BAB61065.1 | |
| AB063301.1 | 2 | 165 | BAB79450.1 | |
| BC028213.1 | AAH28213.1 | |||
| BC038949.1 | 2 | 220 | AAH38949.1 | |
| BG536908.1 | 2 | 274 | ||
| BX397882.2 | 2 | 205 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590102 | Chr.3:39138418 - 39147352 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map