Assay Details
Target Gene Details
Entrez Gene ID: | 81873 |
Gene Name: | actin related protein 2/3 complex subunit 5 like |
Gene Aliases: |
ARC16-2 |
Location: |
Chr.9:124862118-124877733 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPC5L | NM_030978.2 | NP_112240.1 | ||
| XM_011519067.1 | XP_011517369.1 | |||
| AF087842.1 | AAP97155.1 | |||
| AK023881.1 | 1 | 1431 | ||
| AK056174.1 | ||||
| AK315604.1 | ||||
| BC000018.1 | AAH00018.1 | |||
| BC000798.1 | AAH00798.1 | |||
| BC002418.1 | AAH02418.1 | |||
| BX537659.1 | 3 | 5669 |
More Information
Set Membership: |
|

Genomic Map