Assay Details
Target Gene Details
Entrez Gene ID: | 84527 |
Gene Name: | zinc finger protein 559 |
Gene Aliases: |
NBLA00121 |
Location: |
Chr.19:9323772-9343845 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF559 | NM_001202406.1 | 6 | 2174 | NP_001189335.1 |
| NM_001202407.1 | 6 | 2184 | NP_001189336.1 | |
| NM_001202408.1 | 6 | 2252 | NP_001189337.1 | |
| NM_001202409.1 | 7 | 2474 | NP_001189338.1 | |
| NM_001202410.1 | 7 | 2322 | NP_001189339.1 | |
| NM_001202411.1 | 7 | 2351 | NP_001189340.1 | |
| NM_001202412.1 | 7 | 2037 | NP_001189341.1 | |
| NM_032497.2 | 7 | 2237 | NP_115886.1 | |
| AB075488.1 | 2 | 1537 | BAE45747.1 | |
| AK027700.1 | 7 | 1915 | ||
| AK056489.1 | 6 | 1749 | ||
| AK092348.1 | 5 | 2271 | ||
| AK298169.1 | 4 | 1528 | ||
| AK309952.1 | ||||
| AL389937.1 | 1 | 516 | ||
| BC006436.1 | 6 | 2222 | AAH06436.1 |
Target Gene Details
Entrez Gene ID: | 100529215 |
Gene Name: | ZNF559-ZNF177 readthrough |
Gene Aliases: |
- |
Location: |
Chr.19:9324226-9382617 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF559-ZNF177 | NM_001172650.2 | NP_001166121.1 | ||
| NM_001202425.1 | NP_001189354.1 | |||
| U37251.1 | AAB09748.1 |
More Information
Set Membership: |
|

Genomic Map