Assay Details
Target Gene Details
Entrez Gene ID: | 430 |
Gene Name: | achaete-scute family bHLH transcription factor 2 |
Gene Aliases: |
ASH2, HASH2, MASH2, bHLHa45 |
Location: |
Chr.11:2268498-2270952 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASCL2 | NM_005170.2 | NP_005161.1 | ||
| BC057801.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv467645 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain |
|
| nsv951285 | Chr.11:2256071 - 2310670 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map