Assay Details
Target Gene Details
Entrez Gene ID: | 9968 |
Gene Name: | mediator complex subunit 12 |
Gene Aliases: |
ARC240, CAGH45, FGS1, HOPA, MED12S, OHDOX, OKS, OPA1, TNRC11, TRAP230 |
Location: |
Chr.X:71118556-71142454 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED12 | NM_005120.2 | 8 | 1351 | NP_005111.2 |
| AB102668.1 | 5 | 693 | BAC81137.1 | |
| AF071309.1 | 8 | 1164 | AAC83163.1 | |
| AK302543.1 | 8 | 1351 | ||
| D83783.1 | 7 | 993 | BAA12112.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map