Assay Details
Target Gene Details
Entrez Gene ID: | 373509 |
Gene Name: | ubiquitin specific peptidase 50 |
Gene Aliases: |
- |
Location: |
Chr.15:50500562-50546705 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP50 | NM_203494.4 | NP_987090.2 | ||
| AJ583818.2 | CAE47745.2 | |||
| DB448810.2 | ||||
| DB451236.2 | ||||
| DB513975.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv317n27 | Chr.15:50478671 - 50897510 on Build GRCh38 | Gain |
|
| nsv524613 | Chr.15:50364252 - 50576582 on Build GRCh38 | Loss |
|
| nsv1047443 | Chr.15:50452584 - 51354826 on Build GRCh38 | Gain |
|
| nsv569398 | Chr.15:50496928 - 51002798 on Build GRCh38 | Loss |
|
| dgv2594n100 | Chr.15:50469213 - 50823338 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map