Assay Details
Target Gene Details
Entrez Gene ID: | 2327 |
Gene Name: | flavin containing monooxygenase 2 |
Gene Aliases: |
FMO1B1 |
Location: |
Chr.1:171185208-171212683 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FMO2 | NM_001301347.1 | NP_001288276.1 | ||
| NM_001460.4 | NP_001451.2 | |||
| AK304385.1 | ||||
| AK312877.1 | ||||
| AL833218.1 | CAI46191.1 | |||
| BC005894.1 | AAH05894.1 | |||
| BI766311.1 | ||||
| BT006979.1 | AAP35625.1 | |||
| CA312067.1 | ||||
| DB224311.1 | ||||
| DB236882.1 | ||||
| Y09267.1 | CAA70462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28731 | Chr.1:171046945 - 171757821 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map