Assay Details
Target Gene Details
Entrez Gene ID: | 55226 |
Gene Name: | N-acetyltransferase 10 |
Gene Aliases: |
ALP, NET43 |
Location: |
Chr.11:34105564-34146911 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAT10 | NM_001144030.1 | NP_001137502.1 | ||
| NM_024662.2 | NP_078938.2 | |||
| AB051496.1 | ||||
| AF489535.1 | AAO49126.1 | |||
| AK001636.1 | BAA91800.1 | |||
| AK022241.1 | BAB13995.1 | |||
| AK225341.1 | ||||
| AK294044.1 | ||||
| AL136882.1 | CAB66816.1 | |||
| BC020205.1 | ||||
| BC035558.1 | AAH35558.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509400 | Chr.11:34110735 - 34232487 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map