Assay Details
Target Gene Details
Entrez Gene ID: | 79719 |
Gene Name: | alpha- and gamma-adaptin binding protein |
Gene Aliases: |
KPPP1, PPKP1, PPKP1A, p34 |
Location: |
Chr.15:67200675-67255198 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AAGAB | NM_001271885.1 | 10 | 1456 | NP_001258814.1 |
| NM_001271886.1 | 10 | 1717 | NP_001258815.1 | |
| NM_024666.4 | 10 | 1572 | NP_078942.3 | |
| XM_011522020.1 | 10 | 1822 | XP_011520322.1 | |
| AK021568.1 | 10 | 1476 | BAB13845.1 | |
| AL136715.1 | 10 | 1482 | CAB66649.1 | |
| AL832256.1 | 7 | 1153 | CAI46180.1 | |
| BC001975.1 | 7 | 1093 | AAH01975.1 | |
| BC047026.1 | 10 | 1495 | AAH47026.1 | |
| BC058886.1 | 10 | 1494 | AAH58886.1 | |
| BX648272.1 | 1 | 3549 | CAH10544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819309 | Chr.15:67202015 - 67202686 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map