Assay Details
Target Gene Details
Entrez Gene ID: | 100 |
Gene Name: | adenosine deaminase |
Gene Aliases: |
- |
Location: |
Chr.20:44619519-44651758 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADA | NM_000022.3 | 12 | 1500 | NP_000013.2 |
| NM_001322050.1 | 11 | 1384 | NP_001308979.1 | |
| NM_001322051.1 | 11 | 1428 | NP_001308980.1 | |
| NR_136160.1 | 11 | 1435 | ||
| AK123988.1 | 1 | 2558 | ||
| AL832305.1 | 7 | 1715 | ||
| AW450871.1 | 1 | 75 | ||
| BC007678.2 | 12 | 1461 | AAH07678.1 | |
| BC040226.1 | 12 | 1467 | AAH40226.1 | |
| X02994.1 | 12 | 1444 | CAA26734.1 |
More Information
Set Membership: |
|

Genomic Map